

1-(4-Fluorophenyl)cyclopropanecarboxylic acid
Catalog # APOSPC404539-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:180.1800 g/mol
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:773100-29-1
Formula:C₁₀H₉FO₂
MDL Number:MFCD06208777