
2,6-Bis(trifluoromethyl)pyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:455-00-5
:
Formula:C₇H₃F₆N
MDL Number:MFCD00236675
:215.1 g/mol
Boiling Pt:82 °C/18mmHg
Melting Pt:55…58 °C
Flash Pt:67 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together