4-Methyl-6-trifluoromethylpyridin-3-ylamine
Catalog # APOSPC402033-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:176.14 g/mol
Storage Temperature:Ambient
CAS Number:944317-54-8
Formula:C₇H₇F₃N₂
MDL Number:MFCD11848206