Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Registrovat
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(Trifluoromethyl)indoline
4-(Trifluoromethyl)indoline
4-(Trifluoromethyl)indoline
4-(Trifluoromethyl)indoline
APOSPC402024-250MG
:
Apollo Scientific
:
4-(Trifluoromethyl)indoline
APOSPC402024-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
187.1617 g/mol
:
Ambient
:
905274-07-9
:
C₉H₈F₃N
:
MFCD08544326