
3-(1H,1H-Nonafluoropentyl)-4-methylcyclopentane-1,1-dicarboxylic acid
Catalog # APOSPC4009-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:404.23 g/mol
Melting Pt:130…131 °C
:GHS07
Storage Temperature:Ambient
CAS Number:20116-32-9
Formula:C₁₃H₁₃F₉O₄
MDL Number:MFCD01320755