Order Entry
Czech Republic
ContactUsLinkComponent
2-(3,4-Difluorophenyl)ethanol 97%
2-(3,4-Difluorophenyl)ethanol 97%
Catalog # APOSPC400701-5G
Supplier:  Apollo Scientific
CAS Number:  
2-(3,4-Difluorophenyl)ethanol 97%
Catalog # APOSPC400701-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :158.1453 g/mol
Boiling Pt:69 °C/0.1mmHg
Storage Temperature:Ambient
CAS Number:286440-92-4
Formula:C₈H₈F₂O
MDL Number:MFCD00143061