Order Entry
Czech Republic
ContactUsLinkComponent
 
2-[2-(4-Morpholinyl)ethoxy]-5-(trifluoromethyl)aniline
Catalog # APOSPC400019-1G
Supplier:  Apollo Scientific
CAS Number:  
2-[2-(4-Morpholinyl)ethoxy]-5-(trifluoromethyl)aniline
Catalog # APOSPC400019-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :290.29 g/mol
Storage Temperature:Ambient
CAS Number:862873-92-5
Formula:C₁₃H₁₇F₃N₂O₂
MDL Number:MFCD08688414