
2,3,5,6-Tetrafluorobenzonitrile 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:5216-17-1
:
Formula:C₇HF₄N
MDL Number:MFCD00012286
Boiling Pt:88…90 °C/45mmHg
Melting Pt:33…36 °C
Flash Pt:70 °C
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
:175.09 g/mol
:GHS07
Frequently Bought Together