
2'-Fluoro-4'-methoxyacetophenone 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:74457-86-6
:
Formula:C₉H₉FO₂
MDL Number:MFCD00042290
:168.17
Boiling Pt:80…85 °C/0.1mmHg
Melting Pt:51…53 °C
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together