Order Entry
Czech Republic
ContactUsLinkComponent
6-Fluoro-4-hydroxy-2-(trifluoromethyl)quinoline 97%
Irritant
6-Fluoro-4-hydroxy-2-(trifluoromethyl)quinoline 97%
Catalog # APOSPC3756X-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
6-Fluoro-4-hydroxy-2-(trifluoromethyl)quinoline 97%
Catalog # APOSPC3756X-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :231.15 g/mol
Melting Pt:254…260 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:31009-34-4
Formula:C₁₀H₅F₄NO
MDL Number:MFCD00276582