
6-Methyl-4-(trifluoromethyl)-1,6-dihydropyrazolo[3,4-c]pyrazol-3-ylamine 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:478047-15-3
:
Formula:C₆H₆F₃N₅
MDL Number:MFCD02186560
:205.14 g/mol
Melting Pt:122…124 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together