
2,6-Dichloro-α,α,α-trifluorotoluene
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:104359-35-5
:
Formula:C₇H₃Cl₂F₃
Boiling Pt:62…63 °C/9mmHg
Density:0.805 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together