
3-Methoxy-5-(trifluoromethyl)benzoic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:53985-48-1
:
Formula:C₉H₇F₃O₃
MDL Number:MFCD09025409
:220.15 g/mol
Melting Pt:140…142 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together