
2,3-Difluoro-4-hydroxybenzonitrile
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:126162-38-7
:
Formula:C₇H₃F₂NO
MDL Number:MFCD00269596
:155.1 g/mol
Melting Pt:145…149 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together