3-(4-Methoxybenzyloxy)-5-(trifluoromethyl)phenylboronic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:913835-77-5
:
Formula:C₁₅H₁₄BF₃O₄
MDL Number:MFCD08689490
Storage Temperature:Ambient
Frequently Bought Together