Order Entry
Czech Republic
ContactUsLinkComponent
2-Bromo-3-chlorobenzotrifluoride 98%
2-Bromo-3-chlorobenzotrifluoride 98%
Katalogové číslo APOSPC3593-5G
Supplier:  Apollo Scientific
CAS Number:  
2-Bromo-3-chlorobenzotrifluoride 98%
Katalogové číslo APOSPC3593-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :259.46 g/mol
Storage Temperature:Ambient
CAS Number:384-16-7
Formula:C₇H₃BrClF₃
MDL Number:MFCD00673988