2-(Trifluoromethylthio)benzyl bromide 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:238403-52-6
:
Formula:C₈H₆BrF₃S
MDL Number:MFCD00236345
:271.10 g/mol
Boiling Pt:71 °C/1mmHg
Storage Temperature:Ambient
UN:1760
ADR:8,III
:GHS05
Frequently Bought Together