Order Entry
Czech Republic
ContactUsLinkComponent
2-Fluoro-5-(piperidine-1-carbonyl)phenylboronic acid 97%
2-Fluoro-5-(piperidine-1-carbonyl)phenylboronic acid 97%
Catalog # APOSPC3562-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Fluoro-5-(piperidine-1-carbonyl)phenylboronic acid 97%
Catalog # APOSPC3562-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :251.06 g/mol
Storage Temperature:Ambient
CAS Number:874289-43-7
Formula:C₁₂H₁₅BFNO₃
MDL Number:MFCD08436068