
2-Fluoro-(4-methoxycarbonyl)phenylboronic acid 96%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:603122-84-5
:
Formula:C₈H₈BFO₄
MDL Number:MFCD08436034
:197.96 g/mol
Melting Pt:173…175 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together