Order Entry
Czech Republic
ContactUsLinkComponent
8-(2-Fluoro-4-nitrophenyl)-1,4-dioxa-8-azaspiro[4.5]decane
8-(2-Fluoro-4-nitrophenyl)-1,4-dioxa-8-azaspiro[4.5]decane
Catalog # APOSPC3518-1G
Supplier:  Apollo Scientific
CAS Number:  
8-(2-Fluoro-4-nitrophenyl)-1,4-dioxa-8-azaspiro[4.5]decane
Catalog # APOSPC3518-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :282.27 g/mol
Melting Pt:123…125 °C
Storage Temperature:Ambient
CAS Number:252336-82-6
Formula:C₁₃H₁₅FN₂O₄
MDL Number:MFCD08692382