Order Entry
Czech Republic
ContactUsLinkComponent
2,2-Bis(4-isocyanatophenyl)hexafluoropropane 98%
2,2-Bis(4-isocyanatophenyl)hexafluoropropane 98%
Catalog # APOSPC3481-1G
Supplier:  Apollo Scientific
CAS Number:  
2,2-Bis(4-isocyanatophenyl)hexafluoropropane 98%
Catalog # APOSPC3481-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :386.25 g/mol
Storage Temperature:Ambient
CAS Number:10224-18-7
Formula:C₁₇H₈F₆N₂O₂
MDL Number:MFCD00142759