
2-(Trifluoromethoxy)benzenesulphonamide
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:37526-59-3
:
Formula:C₇H₆F₃NO₃S
MDL Number:MFCD01320751
:241.19 g/mol
Melting Pt:187…190 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together