
Diethyl difluoromalonate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:680-65-9
:
Formula:C₇H₁₀F₂O₄
MDL Number:MFCD00221617
:196.15 g/mol
Boiling Pt:94…95 °C/23mmHg
Density:1.179 g/cm³ (25 °C)
Flash Pt:62 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together