
3'-Fluoro-4'-(trifluoromethyl)acetophenone 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:237761-81-8
:
Formula:C₉H₆F₄O
MDL Number:MFCD00236273
:206.14 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together