Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Fluoro-3'-iododiphenyl ether
4-Fluoro-3'-iododiphenyl ether
4-Fluoro-3'-iododiphenyl ether
APOSPC3401-1G
:
Apollo Scientific
:
4-Fluoro-3'-iododiphenyl ether
APOSPC3401-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
314.1 g/mol
:
48…52 °C
:
>110 °C
:
GHS07
:
Refrigerator
:
188534-09-0
:
C₁₂H₈FIO
:
MFCD00236611