
2-(Trifluoromethyl)-3-ethoxydodecafluorohexane 99%
:297730-93-9
:
:C₉H₅F₁₅O
:MFCD04038367
:414.12 g/mol
:130 °C
:–100 °C
:1.61 g/cm³ (25 °C)
:GHS07
:Ambient
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals
Acrylonitrile 99+%
: Thermo Fisher Scientific
Glycerol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 1.000 mL
: PanReac AppliChem
Bis(2-methoxyethyl)ether, extra dry over Molecular Sieve 99+%, AcroSeal®
: Thermo Fisher Scientific
Dimethyl sulphoxide, extra dry over Molecular Sieve 99.7+%, AcroSeal®
: Thermo Fisher Scientific
Glycerine, Hi-LR™
: HIMEDIA
o-Xylene 98%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
: Honeywell Chemicals
Dimethyl sulphoxide, anhydrous ≥99.8%
: Thermo Fisher Scientific
Tris(perfluorobutyl)amine
: Apollo Scientific
Trifluoroacetic acid 99+% for spectroscopy
: Thermo Fisher Scientific
Chloroform ≥99.8% stabilised ACS, Riedel-de Haën™
: Honeywell Chemicals
Diisopropyl ether ≥98.5% (by GC) stabilised, Puriss. p.a., Riedel-de Haën™
: Honeywell Chemicals