
3'-Fluoroacetophenone 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:455-36-7
:
Formula:C₈H₇FO
MDL Number:MFCD00000338
:138.14 g/mol
Boiling Pt:81 °C/9mmHg
Melting Pt:–3 °C
Density:1.126 g/cm³ (25 °C)
Flash Pt:80 °C
EINECS:207-245-4
:GHS07
Storage Temperature:Ambient
Frequently Bought Together