
N'-Hydroxy-2-[4-(trifluoromethyl)phenyl]ethanimidamide
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1312766-79-2
:
Formula:C₉H₉F₃N₂O
MDL Number:MFCD16077266
:218.18 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together