

Ethyl trifluoromethanesulphonate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:425-75-2
:
Formula:C₃H₅F₃O₃S
MDL Number:MFCD00000410
:178.13 g/mol
Boiling Pt:115…118 °C
Density:1.374 g/cm³ (25 °C)
Flash Pt:35 °C
Storage Temperature:Ambient
EINECS:207-037-3
UN:2920
ADR:8+3,II
:GHS02
:GHS05
Frequently Bought Together