Order Entry
Czech Republic
ContactUsLinkComponent
3',5'-Difluoro-4'-methoxyacetophenone 98%
Irritant
3',5'-Difluoro-4'-methoxyacetophenone 98%
Catalog # APOSPC3283-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3',5'-Difluoro-4'-methoxyacetophenone 98%
Catalog # APOSPC3283-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :186.15 g/mol
Melting Pt:27…31 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:170570-79-3
Formula:C₉H₈F₂O₂
MDL Number:MFCD00236243