4,4-Difluorocyclohexanecarbonyl chloride
Catalog # APOSPC32810-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:182.6 g/mol
Storage Temperature:Freezer
CAS Number:376348-75-3
Formula:C₇H₉ClF₂O
MDL Number:MFCD13173983