


4-Fluorophenyl isothiocyanate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1544-68-9
:
Formula:C₇H₄FNS
MDL Number:MFCD00004809
:153.18 g/mol
Boiling Pt:228 °C
Melting Pt:24…28 °C
Density:1.25 g/cm³ (25 °C)
Flash Pt:89 °C
Storage Temperature:Refrigerator
EINECS:216-280-4
UN:1759
ADR:8,II
:GHS05
:GHS07
:GHS08
Frequently Bought Together