
2,4-Difluorothiobenzamide 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:175276-92-3
:
Formula:C₇H₅F₂NS
MDL Number:MFCD00085011
:173.19 g/mol
Melting Pt:128…132 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS07
Frequently Bought Together