
Ethyl-4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carboxylate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:175277-03-9
:
Formula:C₁₄H₁₂F₃NO₂S
MDL Number:MFCD00068104
:315.32 g/mol
Melting Pt:85…86 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together