
Ethyl-4-fluorobenzoate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:451-46-7
:
Formula:C₉H₉FO₂
MDL Number:MFCD00000351
:168.17 g/mol
Boiling Pt:209…210 °C
Melting Pt:25…27 °C
Density:1.146 g/cm³ (25 °C)
Flash Pt:81 °C
EINECS:207-194-8
:GHS07
Storage Temperature:Ambient
Frequently Bought Together