Order Entry
Czech Republic
ContactUsLinkComponent
4'-Chloro-2,2,2-trifluoroacetophenone 97%
4'-Chloro-2,2,2-trifluoroacetophenone 97%
Catalog # APOSPC31735-5G
Supplier:  Apollo Scientific
CAS Number:  
4'-Chloro-2,2,2-trifluoroacetophenone 97%
Catalog # APOSPC31735-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :208.5700 g/mol
Storage Temperature:Ambient
CAS Number:321-37-9
Formula:C₈H₄ClF₃O
MDL Number:MFCD00018686