
4'-(Difluoromethoxy)acetanilide
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:22236-11-9
:
Formula:C₉H₉F₂NO₂
MDL Number:MFCD00467428
:201.1701 g/mol
Melting Pt:114…115 °C
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together