
1H,1H,8H,8H-Dodecafluoro-1,8-octandiol 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:90177-96-1
:
Formula:C₈H₆F₁₂O₂
MDL Number:MFCD00153145
:362.12 g/mol
Boiling Pt:>250 °C
Melting Pt:80…83 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together