1-[2-Nitro-4-(trifluoromethyl)phenyl-[1,4]diazepane hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:646455-48-3
:
Formula:C₁₂H₁₅ClF₃N₃O₂
MDL Number:MFCD09038316
:325.72 g/mol
Storage Temperature:Ambient
Frequently Bought Together