Order Entry
Czech Republic
ContactUsLinkComponent
3,5-Bis(trifluoromethyl)-N-ethylaniline 97%
Irritant
3,5-Bis(trifluoromethyl)-N-ethylaniline 97%
Catalog # APOSPC3107K-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3,5-Bis(trifluoromethyl)-N-ethylaniline 97%
Catalog # APOSPC3107K-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :257.18 g/mol
Boiling Pt:96…98 °C/15mmHg
 :GHS07
Storage Temperature:Ambient
CAS Number:49850-16-0
Formula:C₁₀H₉F₆N
MDL Number:MFCD00203993