α,α,α,α',α',α'-Hexafluoro-m-xylene 99%
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: 1,3-Bis(trifluoromethyl)benzene, α,α,α,β,β,β-Hexafluoro-m-xylene
- CAS Number:402-31-3
- Formula:C₈H₄F₆
- MDL Number:MFCD00000392
- Molecular Weight:214.11 g/mol
- Boiling Pt:115…117 °C
- Melting Pt:–35 °C
- Density:1.378 g/cm³ (25 °C)
- Flash Pt:26 °C
- Storage Temperature:Ambient
- EINECS:206-939-4
- UN:1993
- ADR:3,III
2 Options Available Below
Product Details & Documents
Synonyms: 1,3-Bis(trifluoromethyl)benzene, α,α,α,β,β,β-Hexafluoro-m-xylene
- CAS Number:402-31-3
- Formula:C₈H₄F₆
- MDL Number:MFCD00000392
- Molecular Weight:214.11 g/mol
- Boiling Pt:115…117 °C
- Melting Pt:–35 °C
- Density:1.378 g/cm³ (25 °C)
- Flash Pt:26 °C
- Storage Temperature:Ambient
- EINECS:206-939-4
- UN:1993
- ADR:3,III
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



