
5-(Trifluoromethyl)indole
Catalog # APOSPC3045-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:185.15 g/mol
Melting Pt:67…70 °C
:GHS07
Storage Temperature:Ambient
CAS Number:100846-24-0
Formula:C₉H₆F₃N
MDL Number:MFCD03095341