Order Entry
Czech Republic
ContactUsLinkComponent
4-Ethoxy-2-fluorobenzoic acid 98%
Irritant
4-Ethoxy-2-fluorobenzoic acid 98%
Catalog # APOSPC303114-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Ethoxy-2-fluorobenzoic acid 98%
Catalog # APOSPC303114-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :184.17 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1206593-29-4
Formula:C₉H₉FO₃
MDL Number:MFCD13196593