
4-Fluoro-3-methyl-5-(trifluoromethyl)aniline 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1373920-72-9
:
Formula:C₈H₇F₄N
MDL Number:MFCD22201067
ADR:6.1,III
:193.14 g/mol
UN:2811
:GHS07
Storage Temperature:Ambient
Frequently Bought Together