Order Entry
Czech Republic
ContactUsLinkComponent
(3-Fluoro-2-(trifluoromethyl)phenyl)acetic acid 97%
(3-Fluoro-2-(trifluoromethyl)phenyl)acetic acid 97%
Catalog # APOSPC303068-1G
Supplier:  Apollo Scientific
CAS Number:  
(3-Fluoro-2-(trifluoromethyl)phenyl)acetic acid 97%
Catalog # APOSPC303068-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :222.14 g/mol
Melting Pt:122…124 °C
Storage Temperature:Ambient
CAS Number:897940-14-6
Formula:C₉H₆F₄O₂
MDL Number:MFCD09832251