
2,6-Dichloro-3-(trifluoromethyl)benzylamine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1092461-13-6
:
Formula:C₈H₆Cl₂F₃N
MDL Number:MFCD11519352
:244.04 g/mol
Storage Temperature:Ambient
UN:2735
ADR:8,III
:GHS05
Frequently Bought Together