
3-Methoxy-4-(trifluoromethyl)benzoyl chloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1261571-92-9
:
Formula:C₉H₆ClF₃O₂
MDL Number:MFCD18398587
:238.59 g/mol
Storage Temperature:Ambient
UN:3261
ADR:8,II
:GHS05
Frequently Bought Together