Order Entry
Czech Republic
ContactUsLinkComponent
4-Fluoro-3-methyl-5-(trifluoromethyl)benzoyl chloride 97%
4-Fluoro-3-methyl-5-(trifluoromethyl)benzoyl chloride 97%
Catalog # APOSPC302881-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Fluoro-3-methyl-5-(trifluoromethyl)benzoyl chloride 97%
Catalog # APOSPC302881-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :240.58 g/mol
Storage Temperature:Ambient
CAS Number:1373920-95-6
Formula:C₉H₄ClF₄O
MDL Number:MFCD22201065