Order Entry
Czech Republic
ContactUsLinkComponent
3-Fluoro-2-methoxybenzyl bromide 97%
Corrosive
3-Fluoro-2-methoxybenzyl bromide 97%
Catalog # APOSPC302841-1G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
3-Fluoro-2-methoxybenzyl bromide 97%
Catalog # APOSPC302841-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :219.05 g/mol
Storage Temperature:Ambient
UN:3265
ADR:8,II
 :GHS05
CAS Number:916420-50-3
Formula:C₈H₈BrFO
MDL Number:MFCD09025370