Order Entry
Czech Republic
ContactUsLinkComponent
 
3-[4-Methoxy-3-(trifluoromethoxy)phenyl]propionic acid 97%
Catalog # APOSPC302825-1G
Supplier:  Apollo Scientific
CAS Number:  
3-[4-Methoxy-3-(trifluoromethoxy)phenyl]propionic acid 97%
Catalog # APOSPC302825-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :264.2 g/mol
Storage Temperature:Ambient
CAS Number:1261587-75-0
Formula:C₁₁H₁₁F₃O₄
MDL Number:MFCD18398561